Learn More
Bachem Mating Factor a Trifluoroacetate
SDP
Supplier:  Bachem 4003514.0001
Encompass_Preferred
Mating Factor a Trifluoroacetate, H-Trp-His-Trp-Leu-Gln-Leu-Lys-Pro-Gly-Gln-Pro-Met-Tyr-OH, 59401-28-4, C82H114N20O17S, Mr 1684.01, 1mg. a-Factor, alpha-Factor, Pheromone a-Factor, a-Mating Factor, yeast. Mating pheromone of mating type a cells of S. cerevisiae. This factor inhibits DNA synthesis, thereby preventing cell division of mating type a cells. The hydrophobic peptide binds to a class D G-protein-coupled receptor Ste2p.
Catalog No. 50-382-649