Learn More
Aviva Systems Biology Corp MATN3 Antibody - N-terminal region (ARP56344_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56344_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MATN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MATN3. Alternate names: EDM5, HOA, OS2, DIPOA, OADIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61822