Learn More
Aviva Systems Biology Corp MATN4 Antibody - N-terminal region (ARP65342_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65342_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MATN4. It was validated on Western Blot. . Alternate names: FLJ14417, HE6WCR54ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-480