Learn More
Aviva Systems Biology Corp MATR3 Antibody - C-terminal region (ARP40923_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40923_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MATR3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MATR3. Alternate names: DKFZp686K0542, DKFZp686K23100, KIAA0723, MGC9105, MPD2, VCPDMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53250