Learn More
Aviva Systems Biology Corp MATR3 Antibody - N-terminal region (ARP40922_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40922_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MATR3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MATR3. Alternate names: MPD2, VCPDMApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53249