Learn More
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Specifications
| Product Type | Basement membrane matrix |
| Without Additives | No LDEV |
| For Use With (Application) | Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice |
| Quantity | 100 mL (10 mL x 10) |
| Source | Engelbreth-Holm-Swarm(EHS) Mouse Tumor |
| Content And Storage | Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.