Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Matrigel™ Matrix-3D Plate, 384-well
GREENER_CHOICE

Catalog No. 08774409 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White, Clear
No. per Pack:
1
5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
08-774-409 White, Clear 5
08-774-408 Black 5
08-774-410 White, Clear 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-774-409 Supplier Corning™ Supplier No. 356257
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corning™ Matrigel™ Matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. It has a high-throughput format that can be used for drug discovery with 3D cell cultures such as organoids, spheroids or tumoroids.

  • A high-throughput pre-coated format that contains Matrigel™ and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids
  • Extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors
  • Each lot is verified for intraplate consistency
  • Tested and found negative for the presence of bacteria and fungi
  • Easy and convenient as Matrigel™ is pre-dispensed into each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded 3D application workflows
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart No
Lid With Lid
Content And Storage Store at -20°C
Autoclavable Not Autoclavable
Color White, Clear
Disposable Single-use
Material Polystyrene
No. of Wells 384
Surface Treatment Matrigel Matrix
Well Shape Flat Bottom (F-well)
Dnase Free No
For Use With (Application) 3D cell culture
Includes Guidelines for Use
No. per Case 1
No. per Pack 5
Regulatory Status RUO
Rimmed No
RNase Free No
Skirt Color White with clear bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.