Learn More
Medchemexpress LLC HY-N0164 100mg Medchemexpress, Matrine CAS:519-02-8 Purity:>98%
SDP
Catalog No. 501933438
Encompass_Preferred
Medchemexpress, HY-N0164 100mg Matrine CAS:519-02-8 Matrine (Matridin-15-one) is an alkaloid found in plants from the Sophora genus. It has a variety of pharmacological effects, including anti-cancer effects, and action as a kappa opioid receptor and u-receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3438 Supplier Medchemexpress LLC Supplier No. HYN0164100MG
Medchemexpress, HY-N0164 100mg Matrine CAS:519-02-8 Matrine (Matridin-15-one) is an alkaloid found in plants from the Sophora genus. It has a variety of pharmacological effects, including anti-cancer effects, and action as a kappa opioid receptor and u-receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.