Learn More
Apexbio Technology LLC Matrine(Synonyms: Sophocarpidine, Matridin, (-)-Matrine, α-Matrine, (−)-α-Matrine, L-Matrine, Kushenin), 200mg, CAS: 519-02-8.
SDP
Catalog No. 5000060854
Encompass_Preferred
Matrine (CAS 519-02-8) is an alkaloid derived from plants in the Sophora genus with demonstrated anticancer and anti-inflammatory activities It functions in part through agonism of kappa-opioid and possibly other receptors Matrine inhibits the MNK45 gastric cancer cell line proliferation (IC50 540 g/ml MTT assay) by modifying protein expression profiles (e g NF- B XIAP CIAP p-ERK) In vitro matrine induces apoptosis in NSCLC cells through ROS generation caspase activation and p38 pathway modulation Animal studies suggest therapeutic roles in cardiac injury and sepsis-related lung inflammation
Catalog No. 50-000-60854 Supplier Apexbio Technology LLC Supplier No. A3583200
Matrine (CAS 519-02-8) is an alkaloid derived from plants in the Sophora genus with demonstrated anticancer and anti-inflammatory activities It functions in part through agonism of kappa-opioid and possibly other receptors Matrine inhibits the MNK45 gastric cancer cell line proliferation (IC50 540 g/ml MTT assay) by modifying protein expression profiles (e g NF- B XIAP CIAP p-ERK) In vitro matrine induces apoptosis in NSCLC cells through ROS generation caspase activation and p38 pathway modulation Animal studies suggest therapeutic roles in cardiac injury and sepsis-related lung inflammation