Learn More
TARGETMOL CHEMICALS INC Matrine 200MG
SDP
Catalog No. 502877647
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Matrine (Vegard), an alkaloid isolated from the Sophora genus, acts as a kappa opioid receptor agonist. Purity 97.16%
Catalog No. 50-287-7647 Supplier TARGETMOL CHEMICALS INC Supplier No. T2870200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Matrine (Vegard), an alkaloid isolated from the Sophora genus, acts as a kappa opioid receptor agonist. Purity 97.16%