Learn More
Medchemexpress LLC Matrine | 519-02-8 | 98.0% | 248.37 | 200 MG
SDP
Catalog No. 5000427417
Encompass_Preferred
Matrine (Matridin-15-one) is an alkaloid found in plants of the *Sophora* genus. It acts as an agonist for kappa opioid receptor and u-receptor, and exhibits a range of pharmacological properties, including anti-cancer, anti-oxidative stress, anti-inflammatory, and anti-apoptosis effects. It holds potential for research into diseases such as human non-small cell lung cancer, hepatoma, papillary thyroid cancer, and acute kidney injury (AKI).
  • Acts as a kappa opioid receptor and u-receptor agonist.
  • Demonstrates anti-cancer effects.
  • Possesses anti-oxidative stress properties.
  • Exhibits anti-inflammatory effects.
  • Has anti-apoptosis effects.
  • Potential for research in human non-small cell lung cancer, hepatoma, papillary thyroid cancer, and acute kidney injury (AKI).
Catalog No. 50-004-27417 Supplier Medchemexpress LLC Supplier No. HYN0164200MG
Matrine (Matridin-15-one) is an alkaloid found in plants of the *Sophora* genus. It acts as an agonist for kappa opioid receptor and u-receptor, and exhibits a range of pharmacological properties, including anti-cancer, anti-oxidative stress, anti-inflammatory, and anti-apoptosis effects. It holds potential for research into diseases such as human non-small cell lung cancer, hepatoma, papillary thyroid cancer, and acute kidney injury (AKI).
  • Acts as a kappa opioid receptor and u-receptor agonist.
  • Demonstrates anti-cancer effects.
  • Possesses anti-oxidative stress properties.
  • Exhibits anti-inflammatory effects.
  • Has anti-apoptosis effects.
  • Potential for research in human non-small cell lung cancer, hepatoma, papillary thyroid cancer, and acute kidney injury (AKI).