Learn More
Thermo Scientific Thermo Scientific™ Matrix™ Deepwell Storage Blocks
Catalog No. 50823830 Shop All Thermo Scientific Products
Encompass
ScreenMates 1mL 96 Deep Well Storage Block, Round bottom, Round wells. Polypropylene, Sterile. Can be sealed with Matrix Tech CapMats or Sealing Tape. 40 blocks per case, packaged in Easy Tear bags. Safe for storage down to -80 deg C.
Catalog No. 50-823-830 Supplier Thermo Scientific Supplier No. 421211
ScreenMates 1mL 96 Deep Well Storage Block, Round bottom, Round wells. Polypropylene, Sterile. Can be sealed with Matrix Tech CapMats or Sealing Tape. 40 blocks per case, packaged in Easy Tear bags. Safe for storage down to -80 deg C.