Learn More
Thermo Scientific MATRIX 2ML CAPMATS NS 100/CS
Supplier:  Thermo Scientific 4420
Encompass
96 well square Cap Mats. Non-Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 2mL Deepwell blocks (4221/4222). Suitable for storage down to -80 degrees Celsius. Packaged 100 mats per zip lock bag.
Catalog No. 50-823-886