Learn More
Thermo Scientific Nunc™ 96-Well Cap Mats, Fits Matrix Tech 2mL Deepwell blocks (4221/4222)
Catalog No. 50823888 Shop All Thermo Scientific Products
Encompass
96 well square Cap Mats. Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 2mL Deepwell blocks (4221/4222). Suitable for storage down to -80 degrees Celsius. Packaged 10 mats per Easy Tear bags.
Catalog No. 50-823-888 Supplier Thermo Scientific Supplier No. 4422
96 well square Cap Mats. Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 2mL Deepwell blocks (4221/4222). Suitable for storage down to -80 degrees Celsius. Packaged 10 mats per Easy Tear bags.