Learn More
Thermo Scientific MATRIX 2ML DW BLOCK NS 40/CS
Catalog No. 50823831 Shop All Thermo Scientific Products
Encompass
ScreenMates 2mL 96 Deep Well Storage Block, Round bottom, Square wells. Polypropylene, Non-Sterile. Can be sealed with Matrix Tech CapMats or Sealing Tape. 40 blocks per case, packaged in Easy Tear bags. Safe for storage down to -80 deg C.
Catalog No. 50-823-831 Supplier Thermo Scientific Supplier No. 4221
ScreenMates 2mL 96 Deep Well Storage Block, Round bottom, Square wells. Polypropylene, Non-Sterile. Can be sealed with Matrix Tech CapMats or Sealing Tape. 40 blocks per case, packaged in Easy Tear bags. Safe for storage down to -80 deg C.