Learn More
Thermo Scientific Thermo Scientific™ Matrix™ Deepwell Storage Blocks
Catalog No. 50823832 Shop All Thermo Scientific Products
Encompass
ScreenMates 2mL 96 Deep Well Storage Block, Round bottom, Square wells. Polypropylene, Sterile. Can be sealed with Matrix Tech CapMats or Sealing Tape. 40 blocks per case, packaged in Easy Tear bags. Safe for storage down to -80 deg C.
Catalog No. 50-823-832 Supplier Thermo Scientific Supplier No. 4222
ScreenMates 2mL 96 Deep Well Storage Block, Round bottom, Square wells. Polypropylene, Sterile. Can be sealed with Matrix Tech CapMats or Sealing Tape. 40 blocks per case, packaged in Easy Tear bags. Safe for storage down to -80 deg C.