Learn More
Thermo Scientific Thermo Scientific™ Matrix 96 Well Cap Mat
Catalog No. 50823891 Shop All Thermo Scientific Products
Encompass
96 well Cap Mats. Non-Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 96 well plates, 1.4mL or 0.75mL storage tubes, and 1mL deepwell blocks. Suitable for storage down to -80 degrees Celsius. Packaged 10 mats per Easy Tear bags.
Catalog No. 50-823-891 Supplier Thermo Scientific Supplier No. 443211
96 well Cap Mats. Non-Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 96 well plates, 1.4mL or 0.75mL storage tubes, and 1mL deepwell blocks. Suitable for storage down to -80 degrees Celsius. Packaged 10 mats per Easy Tear bags.