Learn More
Thermo Scientific MATRIX 96W CAPMATS NS 100/CS
Catalog No. 50823876 Shop All Thermo Scientific Products
Encompass
96 well Cap Mats. Non-Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 96 well plates, 1.4mL or 0.75mL storage tubes, and 1mL deepwell blocks. Suitable for storage down to -80 degrees Celsius. Packaged 100 mats per case.
Catalog No. 50-823-876 Supplier Thermo Scientific Supplier No. 4410
96 well Cap Mats. Non-Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 96 well plates, 1.4mL or 0.75mL storage tubes, and 1mL deepwell blocks. Suitable for storage down to -80 degrees Celsius. Packaged 100 mats per case.