Learn More
Thermo Scientific Thermo Scientific™ Matrix 96-Well Cap Mats
Catalog No. 50823880 Shop All Thermo Scientific Products
Encompass
Separable Cap Strips. Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 96 well plates, 1.4mL or 0.75mL storage tubes, and 1mL deepwell blocks. Strip is 1cap by 8 caps. Suitable for storage down to -20 degrees Celsius
Catalog No. 50-823-880 Supplier Thermo Scientific Supplier No. 441411
Separable Cap Strips. Sterile. Made of Ethylene Vinyl Acetate. Fits Matrix Tech 96 well plates, 1.4mL or 0.75mL storage tubes, and 1mL deepwell blocks. Strip is 1cap by 8 caps. Suitable for storage down to -20 degrees Celsius