Learn More
Biomatik Corporation Matrix Metalloproteinase 13 (Mmp13) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU29915
Encompass_Preferred
Matrix Metalloproteinase 13 (Mmp13) Polyclonal Antibody is a Rabbit (Oryctolagus cuniculus) Polyclonal Antibody to Matrix Metalloproteinase 13 (MMP13). The Matrix Metalloproteinase 13 (Mmp13) Polyclonal Antibody has been validated for the following applications: WB. Host: Guinea pig. Species: Rabbit (Oryctolagus cuniculus). Immunogen: Glu70~Thr245. Applications: WB. Target Synonyms: Matrix Metalloproteinase 13 (MMP13). Accession Number: O62806. Gene ID: 100008685. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9209