Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Matrix metalloproteinase-19 | >85.0% | 50 UG
SDP

Catalog No. 5000290968
Encompass_Preferred

Recombinant human MMP-19 protein supplied as a filtered solution for research use. The product contains a partial fragment (L101-G256) expressed in E. coli and is intended for biochemical and extracellular matrix remodeling studies.

  • Species: Human.
  • Sequence: L101-G256 (partial).
  • Expression system: E. coli.
  • Molecular weight: ~18 kDa (reducing SDS-PAGE).
  • Formulation: 0.22 μm filtered solution of 0.1% TFA, 30% CAN.
  • Purity: >85% by reducing SDS-PAGE.
  • Storage: Store at -80°C; avoid repeated freeze-thaw cycles.
  • Available sizes: includes a 50 μg variant.

Catalog No. 50-002-90968 Supplier Medchemexpress LLC Supplier No. HYP7592950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human MMP-19 protein supplied as a filtered solution for research use. The product contains a partial fragment (L101-G256) expressed in E. coli and is intended for biochemical and extracellular matrix remodeling studies.

  • Species: Human.
  • Sequence: L101-G256 (partial).
  • Expression system: E. coli.
  • Molecular weight: ~18 kDa (reducing SDS-PAGE).
  • Formulation: 0.22 μm filtered solution of 0.1% TFA, 30% CAN.
  • Purity: >85% by reducing SDS-PAGE.
  • Storage: Store at -80°C; avoid repeated freeze-thaw cycles.
  • Available sizes: includes a 50 μg variant.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.