Learn More
Enzo Life Sciences Matrix metalloproteinase (MMP) inhibitor profiling kit, fluorometric RED, 96 wells
SDP
Catalog No. NC1752212
Encompass_Preferred
The MMP inhibitor profiling kit, fluorometric (also known as fluorimetric) RED is a complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes, using a quenched fluorogenic substrate.
Catalog No. NC1752212 Supplier Enzo Life Sciences Supplier No. BMLAK3080001
The MMP inhibitor profiling kit, fluorometric (also known as fluorimetric) RED is a complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes, using a quenched fluorogenic substrate.