Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Modifier, 0.2% Pd (2000 ppm Pd), Ricca Chemical
SDP

Catalog No. MMPD2KN500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMPD2KN-500 500 mL Natural Poly Bottle
MMPD2KN-50 50 mL Natural Poly Bottle
MMPD2KN-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MMPD2KN-500 Supplier Ricca Chemical Company Supplier No. MMPD2KN500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural Poly Bottle
CAS 7440-06-4, 7697-37-2, 7440-05-3, 7647-01-0
Concentration or Composition Notes 0.2% Pd,2000ppm Pd
CAS Min % 92.0, 6.16, 0.15, 0.03
CAS Max % 95.0, 6.42, 0.15, 0.03
Physical Form Liquid
Molecular Formula Pt
MDL Number MFCD00011179
Color Brown
Melting Point Approximately 0°C
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.