Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Modifier, 2% (w/v) Ammonium Nitrate in 2% (v/v) HNO3, Ricca Chemical
SDP

Catalog No. MMAN20KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMAN20KN100 100 mL Natural LDPE Bottle
MMAN20KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMAN20KN100 Supplier Ricca Chemical Company Supplier No. MMAN20KN100
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural LDPE Bottle
CAS Min % 93.0, 2.89, 1.93
CAS Max % 97.0, 3.01, 2.0
CAS 7697-37-2, 6484-52-2
Odor Odorless
Color Colorless
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 2% Ammonium Nitrate in 2% HNO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.