Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Modifier, 5% Ni (50,000 ppm Ni), Ricca Chemical
SDP

Catalog No. MMNI50KN50 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMNI50KN-50 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMNI50KN-50 Supplier Ricca Chemical Company Supplier No. MMNI50KN50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural Poly Bottle
CAS 7732-18-5, 7697-37-2, 7440-02-0
Concentration or Composition Notes 5% Ni,50,000ppm Ni
CAS Min % 69.0, 25.82, 3.44
CAS Max % 72.0, 26.88, 3.58
Physical Form Liquid
Color Green
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Type Matrix Modifier
Quantity 50 mL
Percent Purity 0.05
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.