Learn More
Pipette Supplies, Inc. MATRIX PIPETTE BATTERY MC
SDP
Catalog No. 501317647
Encompass
Rechargeable nickel metal hydride (NiMH) batteries for use in 6, 8 and 12 channel Matrix Electronic, Impact and Impact2 multichannel pipettes.Battery must be charged prior to use.Same as 6092-11. Formerly MA 6092, replacement for 6090.
Catalog No. 50-131-7647 Supplier Pipette Supplies, Inc. Supplier No. PS 6092
Rechargeable nickel metal hydride (NiMH) batteries for use in 6, 8 and 12 channel Matrix Electronic, Impact and Impact2 multichannel pipettes.Battery must be charged prior to use.Same as 6092-11. Formerly MA 6092, replacement for 6090.