Learn More
Verichem Laboratories MATRIX PLUS CHOLESTEROL KIT
Catalog No. NC9592196
Encompass_Preferred
Matrix Plus Cholesterol Reference Kit 5 x 15mL. aEc Liquid standards with bovine protein aEc Remarkable optical clarity aEc Contains esterified and free cholesterol from bovine serum aEc Prepared with a known linear relation aEc For calibration or calibration verification aEc Constant protein content, pH and ionic strength across concentration levels aEc Prepared without surfactants, glycol or azide aEc Stable 21 months from date of manufacture. Please note that this is a cold product and ships on ice packs.
This item is not returnable. View return policy
Catalog No. NC9592196 Supplier Verichem Laboratories Supplier No. 9550
Matrix Plus Cholesterol Reference Kit 5 x 15mL. aEc Liquid standards with bovine protein aEc Remarkable optical clarity aEc Contains esterified and free cholesterol from bovine serum aEc Prepared with a known linear relation aEc For calibration or calibration verification aEc Constant protein content, pH and ionic strength across concentration levels aEc Prepared without surfactants, glycol or azide aEc Stable 21 months from date of manufacture. Please note that this is a cold product and ships on ice packs.