Learn More
Thermo Scientific MATRIX PS LIDS W/NTCH ST 80/CS
Catalog No. 50823856 Shop All Thermo Scientific Products
Encompass
Matrix Technologies ScreenMates Microplate Lids, Low-Profile, Polystyrene lid for use with 96 or 384 well microplates, Sterile. Corner notches located at A1, H1 corners. 120 lids per case.
Catalog No. 50-823-856 Supplier Thermo Scientific Supplier No. 432011
Matrix Technologies ScreenMates Microplate Lids, Low-Profile, Polystyrene lid for use with 96 or 384 well microplates, Sterile. Corner notches located at A1, H1 corners. 120 lids per case.