Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ Screw Top Cap Tray Lifter for Capit-All ScrewTop Tube Capper/Decapper

Catalog No. 04100526 Shop All Thermo Scientific Products
Change view
Click to view available options
Compatible Storage Tubes:
0.5 mL Matrix tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Compatible Storage Tubes
04-100-526 0.5 mL Matrix tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-100-526 Supplier Thermo Scientific™ Supplier No. 4090

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Screw Top Cap Tray Lifter

  • Screw Top Cap Tray Lifter for Capit-All ScrewTop Tube Capper/Decapper (4111MAT)
  • Compatibility: 0.5mL Matrix tubes
TRUSTED_SUSTAINABILITY

Specifications

Compatible Storage Tubes 0.5 mL Matrix tubes
Type Screw Top Cap Tray Lifter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.