Learn More
Thermo Scientific MATRIX WM TUBING STD ST 5/CS
Supplier:  Thermo Scientific 20130001
Encompass
Tubing Cartridges; for use with Matrix WellMate; Disposable; Pre-sterilized; Pre-calibrated; Standard Bore; 5 per case, 96 well plate filling and volumes above 20uL
Catalog No. 50-823-760
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.