Learn More
Surmodics Inc MatrixGuard™ Diluent, 50 mL
SDP
Catalog No. NC1553270
Encompass_Preferred
MatrixGuard™ Diluent is a matrix blocking agent that utilizes proprietary components to eliminate heterophilic interferences such as HAMA and RF within immunoassay applications. The product was designed to be used within an existing assay protocol to remove matrix interferences. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies.
Catalog No. NC1553270 Supplier Surmodics Inc Supplier No. SM020050
MatrixGuard™ Diluent is a matrix blocking agent that utilizes proprietary components to eliminate heterophilic interferences such as HAMA and RF within immunoassay applications. The product was designed to be used within an existing assay protocol to remove matrix interferences. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies.