Learn More
Electron Microscopy Sciences Tissue Matrice Spinal Cord Coronal, 0.5 mm Mouse, 2.5 mm W x 32 mm L Cavity, 5 mm Depth
SDP
Catalog No. 501928518
Encompass_Preferred
These spinal cord matrices are precisely machined to ensure reproducible 0.5mm or 1mm sections of mouse or rat spinal cords.
Type: 0.5 mm Mouse
Cavity: 2.5 mm W x 32 mm L
Depth: 5 mm
Catalog No. 50-192-8518 Supplier Electron Microscopy Sciences Supplier No. 69085C
These spinal cord matrices are precisely machined to ensure reproducible 0.5mm or 1mm sections of mouse or rat spinal cords.
Type: 0.5 mm Mouse
Cavity: 2.5 mm W x 32 mm L
Depth: 5 mm