Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Matter - All Mixed Up! A Guided Inquiry
SCI_ED

Catalog No. S02001 Shop All Aldon Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02001 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.
Catalog No. S02001 Supplier Innovating Science™ Supplier No. IS2530
Add to Cart
Add to Cart
WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.

Everything from elements, to molecules, objects, plants, and animals, even you and me, is all made up of matter. Explore the physical properties of matter through the separation of various mixtures.

Everything from elements, to molecules, objects, plants, and animals, even you and me, is all made up of matter. Matter - All Mixed Up! allows students to explore the physical properties of matter through the separation of various mixtures. The included experimental protocol enables students to plan and conduct an experiment on their own with basic guidance and allows the assessment of student learning capabilities.

This kit contains all materials needed for 15 groups to perform the experiment and includes a Teacher's Manual and a Student Answer Packet copymaster.

Specifications

Grade 6+
Class Size 15 Lab Groups
Product Type Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.