Learn More
Ampco Mattock with Fiberglass Handle
SDP
Catalog No. 19174149 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
36 -1/4 in.
Catalog No. Length (English)
19-174-149 36 -1/4 in.
1 options
Catalog No. 19-174-149 Supplier Ampco Supplier No. M55FG
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant mattock. Made in the USA.
  • Please see image for additional information regarding measurements.
  • Head length (a): 15 and 1/4 in.
  • Pick flat head width (b): 3 and 7/8 in.
  • Pick flat head height: 3 in.
  • Length: 36 and 1/4 in.
Specifications
Product Type Mattock
Handle Material Fiberglass
Length (English) 36 -1/4 in.
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov