Learn More
Cayman Chemical Maturation-Inducing Steroid (salmonid) EIA Antiserum, 500 dtn
SDP
Catalog No. 502894290
Encompass_Preferred
A polyclonal antiserum that detects MIS
Catalog No. 50-289-4290 Supplier Cayman Chemical Supplier No. 498502500 DTN
A polyclonal antiserum that detects MIS