Learn More
STEMCELL Technologies STEMdiff™ Midbrain Neuron Maturation Kit, Size: 1 Kit
SDP
Catalog No. 502045474
Encompass
The STEMdiff™ Midbrain Neuron Maturation Kit (Catalog #100-0041) is used in conjunction with the STEMdiff™ Midbrain Neuron Differentiation Kit (Catalog #100-0038) to generate a mixed population of midbrain-type neurons from neural progenitor cells derived from human pluripotent stem cells. These media will produce a population of midbrain neurons (≥ 15% TH-positive dopaminergic neurons; ≥ 90% class III βtubulin-positive neurons; < 10% GFAP-positive astrocytes). Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
Catalog No. 50-204-5474 Supplier STEMCELL Technologies Supplier No. 1000041
The STEMdiff™ Midbrain Neuron Maturation Kit (Catalog #100-0041) is used in conjunction with the STEMdiff™ Midbrain Neuron Differentiation Kit (Catalog #100-0038) to generate a mixed population of midbrain-type neurons from neural progenitor cells derived from human pluripotent stem cells. These media will produce a population of midbrain neurons (≥ 15% TH-positive dopaminergic neurons; ≥ 90% class III βtubulin-positive neurons; < 10% GFAP-positive astrocytes). Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.