Learn More
Medchemexpress LLC Mature TGF beta 1/TG 20ug | 20ug
SDP
Catalog No. 5000193889
Encompass_Preferred
TGF beta 1/TGFB1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines TGF beta 1 is a secreted protein that performs many cellular functions including the control of cell growth cell proliferation cell differentiation apoptosis and can regulate the expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha Mature TGF beta 1/TGFB1 Protein Human (Biotinylated HEK293 Avi) is the recombinant human-derived Mature TGF beta 1/TGFB1 protein expressed by HEK293 with C-Avi labeled tag
Catalog No. 50-001-93889 Supplier Medchemexpress LLC Supplier No. HYP7821320UG
TGF beta 1/TGFB1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines TGF beta 1 is a secreted protein that performs many cellular functions including the control of cell growth cell proliferation cell differentiation apoptosis and can regulate the expression and activation of other growth factors including interferon gamma and tumor necrosis factor alpha Mature TGF beta 1/TGFB1 Protein Human (Biotinylated HEK293 Avi) is the recombinant human-derived Mature TGF beta 1/TGFB1 protein expressed by HEK293 with C-Avi labeled tag