Learn More
Antistat Inc. MAUI S3 White Safety Shoe
Catalog No. 17932128
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-932-128 36
17-932-129 37
17-932-130 38
17-932-131 39
17-932-132 40
17-932-133 41
17-932-134 42
17-932-135 43
17-932-136 44
17-932-137 45
17-932-138 46
17-932-139 47
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 17-932-128 Supplier Antistat Inc. Supplier No. 1071356
Only null left
Add to Cart
Add to Cart
MAUI S3 White Safety Shoe with memory foam insole, energy absorbing heel, and anti-torsion sole is lightweight, durable and completely metal free.
  • Made from Microwash, a highly breatheable microfiber, these shoes are specifically designed for all day comfort
  • Highly effective perspiration and abrasion resistant fabric lining
  • Energy absorbing heel area
  • Memory foam insole for superb comfort, posture support and foor stabilisation
  • Anti-Torsion sole specifically designed to increase stability on irregular and wet surfaces
  • Water and chemical repellent
  • ESD dissipative: conforms to IEC 61340-4-3:2017 (IEC 61340-5-1:2016)
  • Ideal for use within ESD areas, food production and manufacturing, hospitals
Specifications
Color White
Material Microfiber
Size 36
Waterproof Yes
Product Type Controlled Environments Shoes