Learn More
R&D Systems Maurice CE-SDS 25X Internal Standard, 2/pk
SDP
Catalog No. NC1233104
Encompass
Internal standard for addition to each sample for CE-SDS application, 2 vials/pack. For use with CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only.
Catalog No. NC1233104 Supplier R&D Systems Supplier No. 046144/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Internal standard for addition to each sample for CE-SDS application, 2 vials/pack. For use with CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only.