Learn More
R&D Systems Maurice CE-SDS PLUS Application Kit
SDP
Catalog No. NC1610143
Encompass
The CE-SDS PLUS application kit contains all components required to run CE-SDS PLUS application. The kit includes CE-SDS PLUS Cartridges, Separation Matrix, Running Buffer, CE-SDS PLUS Sample Buffer, Wash solution, Conditioning Solutions, Internal Standard, Reagent Vials, Pressure Caps and 96-Well Plates. Molecular weight markers can be ordered separately.
Catalog No. NC1610143 Supplier R&D Systems Supplier No. PSMAK03S/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
The CE-SDS PLUS application kit contains all components required to run CE-SDS PLUS application. The kit includes CE-SDS PLUS Cartridges, Separation Matrix, Running Buffer, CE-SDS PLUS Sample Buffer, Wash solution, Conditioning Solutions, Internal Standard, Reagent Vials, Pressure Caps and 96-Well Plates. Molecular weight markers can be ordered separately.