Learn More
R&D Systems Maurice CE-SDS Separation Matrix, 15 mL
SDP
Catalog No. NC1233408
Encompass
Separation matrix for CE-SDS application, 15 mL. For use with CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only.
Catalog No. NC1233408 Supplier R&D Systems Supplier No. 046386/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Separation matrix for CE-SDS application, 15 mL. For use with CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only.