Learn More
R&D Systems Maurice Sample Vials with Integrated inserts, 0.2 mL, 100/pk
SDP
Catalog No. NC1138265
Encompass
Vials with integrated inserts for samples, accomodates 200 uL sample volume. For use with Maurice, Maurice S., and Maurice C. systems only.
Catalog No. NC1138265 Supplier R&D Systems Supplier No. 046083/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Vials with integrated inserts for samples, accomodates 200 uL sample volume. For use with Maurice, Maurice S., and Maurice C. systems only.