Learn More
Medchemexpress LLC Mauritianin | 109008-28-8 | 5 MG
SDP
Catalog No. 5000421451
Encompass_Preferred
Mauritianin is an orally active kaempferol glycoside isolated from the flowers and leaves of Acalypha indica. It acts as a topoisomerase I inhibitor and possesses hepatoprotective, neuroprotective, and renoprotective activities. This product is intended for research use only.
  • Acts as a topoisomerase I inhibitor
  • Possesses hepatoprotective, neuroprotective, and renoprotective activities
  • Increases vascular tone when combined with Alcesefoliside
  • Stabilizes the DNA-topoisomerase I covalent binary complex
  • Reduced malondialdehyde (MDA) production in CCl4-induced liver damage
  • Preserved reduced glutathione (GSH) levels in CCl4-induced liver damage
  • Maintained ethylmorphine-N-demethylase (EMND) activity in CCl4-induced liver damage
  • Preserved liver, brain, and kidney histoarchitecture
Catalog No. 50-004-21451 Supplier Medchemexpress LLC Supplier No. HYN50385MG
Mauritianin is an orally active kaempferol glycoside isolated from the flowers and leaves of Acalypha indica. It acts as a topoisomerase I inhibitor and possesses hepatoprotective, neuroprotective, and renoprotective activities. This product is intended for research use only.
  • Acts as a topoisomerase I inhibitor
  • Possesses hepatoprotective, neuroprotective, and renoprotective activities
  • Increases vascular tone when combined with Alcesefoliside
  • Stabilizes the DNA-topoisomerase I covalent binary complex
  • Reduced malondialdehyde (MDA) production in CCl4-induced liver damage
  • Preserved reduced glutathione (GSH) levels in CCl4-induced liver damage
  • Maintained ethylmorphine-N-demethylase (EMND) activity in CCl4-induced liver damage
  • Preserved liver, brain, and kidney histoarchitecture