Learn More
Cayman Chemical Maurtnn 10mg
SDP
Catalog No. 502948279
Encompass_Preferred
A flavonoid glycoside and DNA topoisomerase I inhibitor; stabilizes the covalent complex between DNA and recombinant human topoisomerase I at 100 µM
Catalog No. 50-294-8279 Supplier Cayman Chemical Supplier No. 3432610 MG
A flavonoid glycoside and DNA topoisomerase I inhibitor; stabilizes the covalent complex between DNA and recombinant human topoisomerase I at 100 µM