Learn More
Medchemexpress LLC HY-109037 10mg Medchemexpress, Mavacamten CAS:1642288-47-8 Purity:>98%
SDP
Catalog No. 501968655
Encompass_Preferred
Medchemexpress, HY-109037 10mg Mavacamten CAS:1642288-47-8 Mavacamten is a modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8655 Supplier Medchemexpress LLC Supplier No. HY10903710MG
Medchemexpress, HY-109037 10mg Mavacamten CAS:1642288-47-8 Mavacamten is a modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.