Learn More
Medchemexpress LLC Mavacamten-d5 | 2453251-00-6 | 98.6% | 278.36 | 1 MG
SDP
Catalog No. 5000346628
Encompass_Preferred
Mavacamten-d5 is a deuterium-labeled version of Mavacamten (HY-109037), an orally active modulator of cardiac myosin. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-46628 Supplier Medchemexpress LLC Supplier No. HY109037S31MG
Mavacamten-d5 is a deuterium-labeled version of Mavacamten (HY-109037), an orally active modulator of cardiac myosin. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.