Learn More
Medchemexpress LLC Mavacamten-d5 | 2453251-00-6 | 98.6% | 278.36 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY109037S310MG
Encompass_Preferred
Mavacamten-d5 is a deuterium labeled Mavacamten, an orally active modulator of cardiac myosin. It has IC50s of 490 nM for bovine cardiac and 711 nM for human cardiac.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-45147