Learn More
Medchemexpress LLC Mavacamten-d5 | 2453251-00-6 | 98.6% | 278.36 | C15H14D5N3O2 | 5 MG
SDP
Catalog No. 5000264306
Encompass_Preferred
Mavacamten-d5 is a deuterium-labeled isotopologue of mavacamten supplied for research use as a stable-isotope reference and tracer in analytical and mechanistic studies. Supplied as a 5 mg solid, the compound has molecular formula C15H14D5N3O2, molecular weight 278.36, and reported HPLC purity 98.61% with isotopic enrichment of 100%.
  • Deuterium-labeled internal standard for mass spectrometry and LC-MS assays.
  • High isotopic enrichment for accurate quantitation and tracer studies.
  • High HPLC purity for consistent analytical performance.
  • Small, well-characterized solid form for convenient handling and storage.
  • Appropriate for method development, pharmacokinetic, and mechanistic research applications.
Catalog No. 50-002-64306 Supplier Medchemexpress LLC Supplier No. HY109037S35MG
Mavacamten-d5 is a deuterium-labeled isotopologue of mavacamten supplied for research use as a stable-isotope reference and tracer in analytical and mechanistic studies. Supplied as a 5 mg solid, the compound has molecular formula C15H14D5N3O2, molecular weight 278.36, and reported HPLC purity 98.61% with isotopic enrichment of 100%.
  • Deuterium-labeled internal standard for mass spectrometry and LC-MS assays.
  • High isotopic enrichment for accurate quantitation and tracer studies.
  • High HPLC purity for consistent analytical performance.
  • Small, well-characterized solid form for convenient handling and storage.
  • Appropriate for method development, pharmacokinetic, and mechanistic research applications.