Learn More
Medchemexpress LLC Mavacamten-d7 | 1642288-47-8 | 99.7% | 280.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY109037S41MG
Encompass_Preferred
Mavacamten-d7 (MYK461-d7) is a deuterium labeled variant of Mavacamten. Mavacamten is an orally active modulator of cardiac myosin, with IC50s of 490 nM and 711 nM for bovine cardiac and human cardiac, respectively.
  • Deuterium labeled compound
  • Modulator of cardiac myosin
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-45723