Learn More
Medchemexpress LLC HY-119447 5mg Medchemexpress, Mavacoxib CAS:170569-88-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1194475MG
Encompass_Preferred
Medchemexpress, HY-119447 5mg Mavacoxib CAS:170569-88-7 Mavacoxib is a selective, oral long-acting cyclooxygenase-2 (COX-2) inhibitor and a long-acting non-steroidal anti-inflammatory drug (NSAID). Mavacoxib is used to treat pain and inflammation associated with degenerative joint disease in dogs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9537